Joulemètre avec couvercle
Lieferant: BREUKHOVEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
27911
761-0049EA 0 EUR InStock
761-0049
Joulemètre avec couvercle
Lehrmittel Physik-Lehrmittel Heat/Thermodynamics Learning Activities
For determining the specific heat of solids and liquids. Aluminium interior, plastic exterior. Plastic cover with slot for thermometer, stirring rod, and immersion thermostat of 12 ohms with a 4 mm connector.
  • Voltage: 10 V
  • Capacity: 200 ml
Height: 130 mm, Diameter: 95 mm
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge