Ball and ring, vans Gravesande
Lieferant: BREUKHOVEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31220
761-0064EA 0 EUR InStock
761-0064
Ball and ring, vans Gravesande
Lehrmittel Physik-Lehrmittel Heat/Thermodynamics Learning Activities
Ball diameter: 25 mm, ring diameter: 25 mm.
  • Length: 190 mm
Rod with ring.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge