Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Skeleton model, Stan

Lieferant: Binhold, Paul Lehrmittel

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

1020171
763-0003EA 852 EUR InStock
763-0003
Skeleton model, Stan
Modelle Humanbiologie
Stan, the standard model of a human skeleton, has been appreciated throughout the world for decades. Thanks to its very high quality and robust construction, it is perfect for use in hospitals, schools, universities and laboratories. Available on a stable metal stand with five casters. Model comes with metal stand and transparent dust cover.

  • Final assembly carried out by hand
  • Made from a durable, unbreakable synthetic material
  • On a stable metal stand with five casters (painted white)
  • Close to the realistic weight of around 200 bones
  • Natural skeleton size
  • Three part assembled skull
  • Individually inserted teeth
  • Limbs can be removed quickly and easily

Dimensions: 176,5 cm

Weight: 9,57 kg

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge