Heart / lung model, 5 parts
Lieferant: Binhold, Paul Lehrmittel
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1001243
763-0169EA 710 EUR InStock
763-0169
Heart / lung model, 5 parts
Modelle Humanbiologie
Model mounted on plate in life size. Shows the larynx, esophagus and trachea, heart, vena cava, aorta, large carotid artery, lungs and diaphragm.
  • Dimensions 12 × 28 × 37 cm
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge