Mini molecular models: DNA
Lieferant: SPIRING ENTERPRISES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MDNA-060-11 MDNA-060-22
763-0453EA 0 EUR InStock
763-0453 763-0454
Mini molecular models: DNA
Modelle Molekularbiologie
Mini molecular models of DNA with nucleic acids, five-sided sugar molecules and pyramid-shaped phosphate molecules.
Only correct base pairs can be connected.
Bestellinformation: Components include colour-coded bases: G, C, A, T.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge