RNA model
Lieferant: SPIRING ENTERPRISES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MRNA-12
763-0455EA 0 EUR InStock
763-0455
RNA model
Modelle Molekularbiologie
RNA 12 base single stranded model kit.
  • Two TRNA and two amino acid parts
  • Original, unique and dual-scale system
  • Can be mounted on the DNA-model
These sets are suitable for students but may also be used by scientists.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge