VWR®, Sterile Alliance Sleeves
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
76491-706EA 477 EUR InStock
76491-706
VWR®, Sterile Alliance Sleeves
Einweg-Kleidung
VWR® Made in USA sterile sleeves are an ideal option for single-use needs. Made with the specially formulated alliance breathable microporous fabric.
  • Individually bagged
  • Available with thumb loop configuration
  • ETO sterilised
  • Made with alliance breathable microporous fabric
  • Made in USA
A key part of the VWR® Made in USA PPE line, these sterile cleanroom sleeves provide a comfortable fit while meeting sterility and filtration requirements of many controlled environments.
The Alliance fabric is designed to lower particulate levels by providing extremely smooth outer fabric surface that lowers particle cling and build-up in cleanroom applications. Alliance products have a unique antistatic system that assures static dissipation in sensitive cleanroom applications, while providing a liquid-proof barrier that ensures protection from hazardous substances used in biotech and pharmaceutical cleanrooms.
Verpackung: Bagged 1 pair per sterile bag, 100 pairs per case. Packed in poly-lined bag in brown corrugated box.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge