Thomson e / m tube
Lieferant: Binhold, Paul Lehrmittel
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1000617
765-0090EA 2430 EUR InStock
765-0090
Thomson e / m tube
Lehrmittel Physik-Lehrmittel Heat/Thermodynamics Learning Activities
Evacuated glass flask containing i.a. electron gun with directly heated tungsten wire cathode and a cylindrical, collimating anode. The generated beam is allowed to stroke along a semi-transparent mica disc in the center of the tube. Luminescent coating on the front of the disc makes the beam appear. Two of the edges of the back are provided with graduated scales.
  • Cut to any length "
The tube can be used for electrostatic and magnetic deflection of the electron beam (the latter with the help of Helmholtz coils), determination of e / m with combined electric and magnetic fields or with only magnetic deflection, which gives a more accurate value. Furthermore for demonstration of the electrons' speed distribution, reflection, spiralization and focusing as well as beam divergence with magnetic alternating field.
Maximum glow voltage: 7.5 V
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge