Communicating tubes
Lieferant: GLASWARENFABRIK KARL HECHT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
41954010
765-0969EA 59.8 EUR InStock
765-0969
Communicating tubes
Lehrmittel Physik-Lehrmittel Mechanics Learning Activities
Five glass tubes of different shape and inner dimensions. Down to each other. When filling the liquid in the pipes, it is clear that the height of the stowage in the tubes. pipes are independent of the design of the pipes. Comes with foot.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge