Demonstration thermometer
Lieferant: MOLLER THERM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
101026
765-1079EA 16.9 EUR InStock
765-1079
Demonstration thermometer
Lehrmittel Physik-Lehrmittel Heat/Thermodynamics Learning Activities
Milk glass scale on polished wooden board. Colored liquid filling. Measuring range -10 to +100 ° C, 0 to +80 ° R, +20 to +212 ° F.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge