Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Radiometer

Lieferant: MOLLER THERM

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

807519
765-1092EA 0 EUR InStock
765-1092
Radiometer
Lehrmittel Physik-Lehrmittel Heat/Thermodynamics Learning Activities
Radiometer according to Crookés. Consists of a glass container on foot with low air pressure and needle-bearing spinner with four vertical mica discs. The discs are black on one side and shiny on the other. The discs thus absorb radiation differently. The higher temperature of the dark side means that the air molecules get greater kinetic energy in collision with the dark side of the discs than with their shiny side. The result is that the spinner starts to rotate.

  • Height: 200 mm
  • Diam: 70 mm

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge