Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Dextrose, wasserfrei 97,5-102,0% (durch Trockensubstanz), körnig, GenAR® USP, Macron Fine Chemicals™

Lieferant: MACRON AVANTOR BRAND

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

7730-06EA 256.18 EUR InStock
7730-06 7730-88
Dextrose, wasserfrei 97,5-102,0% (durch Trockensubstanz), körnig, GenAR® USP, Macron Fine Chemicals™
D(+)-Glucose


Formel: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146
Dichte: 1.54 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00148912
CAS-Nummer: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=18aebec8588c01e468167161dd721370b55777b1&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications
GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C: Water (H₂O) ≤1.0 %
USP - Assay (Anhydrous basis) 97.5 - 102.0 %
USP - Related substances: Maltose and isomaltose ≤0.4 %
USP - Dextrin Passes Test
USP - Related substances: Maltotriose ≤0.2 %
USP - Related substances: Fructose ≤0.15 %
USP - Related substances: Unspecified impurities, each ≤0.10 %
USP - Related substances: Total Impurities ≤0.5 %
USP - Color and Clarity of Solution Passes Test
USP - Soluble Starch, Sulfites ≤15 ppm
USP - Conductivity (uS cm-1) ≤20 µS/cm
EP/BP - Identification A (Specific Rotation (+)) 52.5 - 53.3 Degree
EP/BP - Identification B Passes Test
EP/BP - Identification E Passes Test
EP/BP - Assay (C₆H₁₂O₆) (anhydrous substance) 97.5 - 102.0 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Conductivity, uS cm-1 ≤20
EP/BP - Related Substances: sum of impurities A and B ≤0.4 %
EP/BP - Related Substances: impurity C ≤0.2 %
EP/BP - Related Substances: impurity D ≤0.15 %
EP/BP - Related Substances: unspecified impurities, each ≤0.10 %
EP/BP - Related Substances: total impurities ≤0.5 %
EP/BP - Dextrins Passes Test
EP/BP - Soluble starch, sulfite ≤15 ppm
EP/BP - Water (H₂O) ≤1.0 %
Endotoxin Concentration (EU/g) ≤5.0
JP - Identification 1 Passes Test
JP - Identification 2 Passes Test
JP - Water (H₂O) ≤1.0 %
JP - Assay (Anhydrous basis) 97.5 - 102.0 %
JP - Maltose and isomaltose ≤0.4 %
JP - Maltotriose ≤0.2 %
JP - Fructose ≤0.15 %
JP - Unspecified Impurities, each ≤0.10 %
JP - Total Impurities ≤0.5 %
JP - Color and Clarity of Solution Passes Test
JP - Soluble Starch, Sulfites ≤15 ppm
JP - Conductivity (uS cm-1) ≤20 µS/cm
JP - Dextrin Passes Test
JP - Heavy Metals (as Pb) ≤4 ppm
Appearance (white, granular powder)
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.

 

PRODUKT

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge