Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Natriummetabisulfit 65,0 - 67,4%, körnig NF, FCC, Macron Fine Chemicals™

Lieferant: MACRON AVANTOR BRAND

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

7776-26EA 3098.46 EUR InStock
7776-26 MACR7776-06
Natriummetabisulfit 65,0 - 67,4%, körnig NF, FCC, Macron Fine Chemicals™
Natriummetabisulfit


Formel: Na₂S₂O₅
MW: 190.11 g/mol
Melting Pt: 150
Storage Temperature: Raumtemperatur
MDL Number: MFCD00167602
CAS-Nummer: 7681-57-4
EINECS: 231-673-0
Merck Index: 13,08712
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Food GMP Manufactured Product
Meets N.F. Requirements
GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
NF - Assay (SO₂) 65.0 - 67.4 %
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Thiosulfate (S₂O₃) ≤0.05 %
NF - Iron (Fe) ≤0.002 %
NF - Chloride (Cl) ≤0.05 %
FCC - Assay (Na₂S₂O₅) 96.5 - 100.0 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Iron (Fe) ≤0.002 %
FCC - Lead (Pb) ≤2 mg/kg
FCC - Selenium (Se) ≤5 mg/kg
Solubility (10 in 100) Passes Test
Appearance (white or yellowish white crystalline granules)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge