Protein blocking agent, BLOT-QuickBlocker™
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-011
786-011EA 101 EUR InStock
786-011
Protein blocking agent, BLOT-QuickBlocker™
Puffer Elektrophorese-Puffer Blockier-Puffer
BLOT-QuickBlocker™ is a modified milk protein that is highly soluble and does not inhibit peroxidase detection. The modified milk protein has high blocking efficiency with a clear background.
  • Readily soluble and produces semi-clear solution
  • No inhibition to peroxidase
  • Fat free and produces clear background
  • Higher blocking efficiency
  • Blocking time 30 to 60 minutes
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge