Device for protein cross linking, coupling and modification, Tube-O-Reactor™
Lieferant: G-Biosciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-027-15K 786-027-4K 786-027-8K 786-024-15K 786-024-4K 786-024-8K
786-027-15KEA 253 EUR InStock
786-027-15K 786-027-4K 786-027-8K 786-024-15K 786-024-4K 786-024-8K
Device for protein cross linking, coupling and modification, Tube-O-Reactor™
Röhrchen Dialyseröhrchen
Tube‐O‐Reactor™, Micro, for 20 to 250 µl samples or Medi for 0,2 to 2 ml samples.
The Tube‐O‐Reactor™ is a simple reactor-device for performing a wide variety of reactions including crosslinking, modification, and labeling of proteins and nucleic acids. Most reactions involve three critical steps ‐ equilibration of reaction conditions for efficient reaction, reaction with target specific agents, and then removal of un-reacted agents and bi‐products, followed by recovery of the reaction products. Tube‐O-Reactor™ has been designed to perform all three critical steps of any such reaction with minimum user interventions or hands‐on efforts, which uses only an ordinary laboratory shaker.
The Tube‐O‐Reactor™ consists of a sealed reaction chamber box provided with Tube‐O-Dialyzer™ and dialysis assemblies (includes micro-dialysis cups, floats and stirring balls) in one kit. Each Tube‐O-Reactor™ is suitable for 5 separate reactions, per sample volume of Micro or Medi size. Additional Tube‐O‐Dialyzer™ can be purchased separately. 
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge