Protease assay kit
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-028
786-028EA 697 EUR InStock
786-028
Protease assay kit
Tests Enzymtests
This Protease Assay Kit is designed for the quantitative determination of proteases present in protein sample, using a dye-labelled protein substrate. The proteases present in the sample of interest will digest the protein substrate and release dye labelled peptides. The absorbance of the dye-labelled peptide is measured at 570 nm for determination of protease activity.
Chemically stabilised Trypsin (MSG-Trypsin™) is supplied with the kit as a general protease standard; however, other specific protease standards can also be used. MSG-Trypsin™ is an ultra-pure trypsin from bovine pancreas, modified by methylation followed by TPCK treatment and is extremely resistant to autolysis. The kit components are sufficient for 50 assays in a microtitre plate format or 0,5 ml assay tubes.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge