Pepstatin A
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-052
786-052EA 492 EUR InStock
786-052
Pepstatin A
Proteine und Peptide
Pepstatin is a potent inhibitor of various aspartic proteases, including cathepsin D, renin, pepsin, bacterial aspartic proteases and HIV proteases. Soluble in DMSO or methanol.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge