ELISA kits, femtoELISA™
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-110 786-111 786-112 786-113
786-110EA 668 EUR InStock
786-110 786-111 786-112 786-113
ELISA kits, femtoELISA™
Tests ELISAs
The femtoELISA™ kits utilise a non animal protein blocker, NAP-BLOCKER™ that minimises cross-reactivity with researcher's antigens and antibodies. For the detection of HRP, an improved, ultra sensitive, non volatile, stable colorimetric substrate based on tetramethyl benzidine (TMB) is supplied. Reagent does not require hydrogen peroxide that can have detrimental effects on many biological factors, which may ultimately affect the assays.
For the detection of alkaline phosphatase, a p-nitrophenylphosphate (pNPP) based substrate is supplied that has superior stability compared to commonly used pNPP tablets and solutions. The improved stability ensures minimal background Ø over longer periods compared to normal pNPP substrates. Our highly rapid AP substrate has superior sensitivity and requires no preparation time.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge