Sample clean up, Perfect-FOCUS™
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-124T 786-124
786-124TEA 101 EUR InStock
786-124T 786-124
Sample clean up, Perfect-FOCUS™
Elektrophorese-Reagenzien Western Blotting Reagents
Perfect-FOCUS™ is based on quantitative precipitation and concentration of protein solutions using UPPA™ (Universal Protein Precipitation Agent) and has been specifically developed for preparing lower conductivity protein samples for iso-electric focusing gels. Perfect-FOCUS™ is designed to clean and remove interfering agents from protein samples and concentrate protein solutions that give poor protein spot resolution during 2D electrophoresis.
Protein solutions as dilute as 1 ng/ml, if treated with Perfect-FOCUS™, can be quantitatively precipitated into a small volume. Protein precipitation is not affected by the presence of detergents or other common laboratory agents.
The collected protein sample has a conductivity of approximately 40 to 50 µS and is substantially free from non-protein agents, making the protein sample ideal for critical IEF and 2D gel electrophoresis studies, with improved spot resolution and greatly reduced spot streaking. Perfect-FOCUS™ treated protein samples are compatible with mass spectrometry analysis and show identical peptide spectra.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge