Protein concentration kit, OrgoSol-PROTEIN-Concentrate™
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-125
786-125EA 340 EUR InStock
786-125
Protein concentration kit, OrgoSol-PROTEIN-Concentrate™
Protein-Extraktion Protein Extraction Kits
The OrgoSol-PROTEIN-Concentration™ Kit precipitates protein with a proprietary solvent buffer, OrgoSol™. The OrgoSol™ buffer has been specifically developed for efficient precipitation of protein solutions with minimal disruption to the protein structure and therefore maintains the biological activity of most proteins.
The kit has been extensively tested for the concentration of a wide selection of enzymatic proteins without the loss of their biological activity and for ~100% protein recovery. The kit is designed to precipitate up to 5 ml protein solution.
The method involves mixing a protein solution with the OrgoSol™ Buffer followed by incubation, which results in quantitative precipitation of the protein. The precipitated protein is suspended in a smaller volume of an appropriate buffer and the concentrated protein solution is ready for use.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge