Arrest™ extraction buffer
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-133
786-133EA 410 EUR InStock
786-133
Arrest™ extraction buffer
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
Arrest™ Extraction Buffer is an optimised combination of various chaotropic agents and RNase inhibitors, which inhibits RNase rapidly. It may be used in conjunction with any RNA extraction method, including extractions based on phenol, chloroform and other organic solvents and detergents.
  • Rapidly inhibit destructive RNases
  • Strong chaotropic buffer for rapid RNA release
  • Compatible with numerous species and tissues
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge