Yeast Geno-DNA Template™
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-134
786-134EA 551 EUR InStock
786-134
Yeast Geno-DNA Template™
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
Yeast Geno-DNA Template™ isolates high quality genomic DNA from yeast. The kit is based on a two-step lysis of yeast cells, using LongLife™ Zymolyase® and LongLife™ Proteinase K, followed by the removal of proteins and other cellular impurities by precipitation and centrifugation.
The genomic DNA is collected by precipitation and the high yield of extracted DNA has an average 100 kb length and has A₂₆₀/A₂₈₀ 1,8 to 2,0.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge