Genomic Tube-O-Dialyzer™
Lieferant: G-Biosciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-142-45MC
786-142-45MCEA 510 EUR InStock
786-142-45MC
Genomic Tube-O-Dialyzer™
Röhrchen Dialyseröhrchen
Genomic Tube‐O‐DIALYZER™ (0.45µm). 
Genomic Tube‐O‐DIALYZER™ allows dialysis of small genomic-DNA samples without taking the sample out of the tube and eliminates loss. A simple design converts a sample tube into a dialyzer– Genomic Tube‐O-DIALYZER™ in which the tube cap is adapted with a dialysis membrane. Invert the Genomic‐Tube‐O‐DIALYZER™ in a dialysis tank and dialyze your samples. Salts and other molecules rapidly exchange across the dialysis membrane mounted on the cap. When dialysis is complete, spin the tube for a brief 5 seconds and collect 100% of your sample. Sample collection by spinning allows clean and 100% recovery. There is no left over sample as in a dialysis bags. After dialysis, replace the dialyzing cap with a Tube‐O‐Dialyzer™ Cap (MEDI) supplied with each kit for storage of the sample. Genomic Tube‐O‐DIALYZER™ is supplied with 0.45µm size membrane, which allows rapid removal of impurities, RNA, small fragment DNA etc. and retains only genomic DNA.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge