Protein extraction kit, tissue PE LB™
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-181 786-181T
786-181EA 298 EUR InStock
786-181 786-181T
Protein extraction kit, tissue PE LB™
Puffer Extraktionspuffer
Tissue PE LB™ has been developed for extraction of total biologically active, soluble proteins from animal tissues. Tissue PE LB™ is based on an organic buffer and utilises a mild non-ionic detergent, chelating agent, and a proprietary combination of various salts and agents to enhance extraction and stability of proteins. Depending on the required downstream application, additional agents such as reducing agents and protease inhibitors may be added into Tissue PE LB™. Tissue PE LB™ has been tested for use with a wide variety of fresh and frozen animal tissues.
  • Provides a simple and versatile method for protein extraction from fresh or frozen tissues
  • Compatible with electrophoresis and chromatographic applications
  • Compatible with most enzyme assays including: Reporter gene assays (ß-galactosidase, luciferase, chloramphenicol acetyltransferase); Kinases (protein kinase C, protein kinase A, tyrosine kinase); Immunoassays (ELISA, Western blots, RIA)
  • Suitable for most applications including enzyme and protein purification applications, electrophoresis, Western blotting and 2D-gel analysis
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge