CytoScan™ Cell proliferation assay kit, WST-1
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-212
786-212EA 582 EUR InStock
786-212
CytoScan™ Cell proliferation assay kit, WST-1
Tests Zelltests
CytoScan™ WST-1 Cell Proliferation Assay principle is based upon the reduction of the tetrazolium salt WST-1 to formazan by cellular dehydrogenases. The generation of the dark yellow coloured formazan is measured at 420 to 480 nm (optimal at 440 nm) and is directly correlated to cell number.
  • Sensitive and accurate assay
  • Colorimetric assay
  • No washing, harvesting or solubilization required
  • For the detection of cell toxicity, death, viability or proliferation
  • Ideal for high throughput screening
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge