InGel™ Array kit for protein digestion
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-241A
786-241AEA 1240 EUR InStock
786-241A
InGel™ Array kit for protein digestion
Protein-Extraktion Protein Extraction Kits
InGel™ Array is a 96-well format kit suitable for processing larger numbers of protein spots concurrently and is compatible with spot-picking instruments. Fewer than 96 samples a batch may also be processed without any waste.
  • Digestion of protein spots in 96 well titre plate format
  • Suitable for 2D gels stained with silver stain, Coomassie® dye, and other methods
The protein spots are first excised from the silver stained gel and transferred to a proteomic grade titre plate. Silver stained gel pieces are washed with SilverOUT™ to remove inhibitory silver ions. The proteins are then digested within the gel using a mass spectrometry grade trypsin and a supplied trypsin digestion buffer.
The digested peptides are extracted with G-Biosciences' Pep-Extract™, a high diffusion peptide extraction buffer. The extracted peptides are suitable for mass spectrometry analysis without any subsequent treatments or cleaning procedures.
Lieferumfang: InGel™ Array kit is supplied with reagents sufficient for up to 500 protein spots. The kit includes SilverOUT™ for the removal of silver ions, trypsin digestion buffer for optimal trypsin activity, Pep-Extract™ for high level peptide extraction, and the InGel™ Array titre plates and caps. MSG-Trypsin™ (Mass Spectrometry Grade) trypsin, which increases stability, is also available but sold separately.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge