Protein purification kit, thiophilic adsorption
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-268 786-266 786-267
786-268EA 3070 EUR InStock
786-268 786-266 786-267
Protein purification kit, thiophilic adsorption
Protein-Aufbereitungs Protein Purification Kits
Thiophilic adsorption or thiophilic chromatography is a routinely used technique for the low cost, simple purification of immunoglobulins. Thiophilic adsorption was first developed by Porath et al in 1984 and is a group specific, salt-dependent purification technique that has distinct affinity towards immunoglobulins and α2- macroglobulins. The thiophilic adsorption works on the principle that some proteins in high salt are able to bind to an immobilised ligand that contains a sulfone group in proximity to a thioether group. The bound proteins are then eluted in decreasing salt concentrations. The thiophilic resin binds immunoglobulins, including IgG, IgY and IgM, from serum, ascites or tissue culture supernatants and the purified immunoglobulins are then eluted in a near neutral aqueous buffer.
The thiophilic resin has a high binding capacity (~20 mg/ml human IgG/ml resin) and a broad specificity for various immunoglobulin molecules. Thiophilic adsorption has been used to purify other proteins including horseradish peroxidase, glutathione peroxidase, lactate dehydrogenase and allergens.
Lieferumfang: Supplied with protocols for IgG purification, IgM purification, IgY purification and general protein purification. The Thiophilic Adsorption kit is supplied with the thiophilic resin and all the necessary buffers for the rapid purification of immunoglobulin G (IgG) antibodies.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge