FASTsilver™, rapid silver stain for protein gels and nucleic acids
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-30
786-30EA 430 EUR InStock
786-30
FASTsilver™, rapid silver stain for protein gels and nucleic acids
Färbemittel Elektrophorese-Färbemittel
G-Biosciences' FASTsilver™ is a rapid silver stain for nucleic acids and proteins in electrophoresis gels
  • Nanogram-sensitive
  • Stains both nucleic acids & proteins
  • Maximum visibility: produces crystal clear backgrounds & sharp protein bands
  • Lower Detection Limit: 1ng/band
FASTsilver™ does not use the protein modifier glutaraldehyde, and therefore, it allows for complete recovery of proteins and trypsin digested peptides.
G-Biosciences' FASTsilver™ detects as little as 0.3 ng nucleic acid and 1 ng BSA protein. The kit contains ready-to-use reagents for 25 mini-gels and comes with a simple-to-follow 60 to 90 minute protocol.
The FASTsilver™ kit is a nanogram-sensitive silver staining kit that produces crystal clear background and maximal sensitivity needed for critical analysis. This unique formulation of reagents leaves the background clear and produces sharp images of nucleic acid bands.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..