Cell surface protein isolation and labelling kit, HOOK™
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-316
786-316EA 666 EUR InStock
786-316
Cell surface protein isolation and labelling kit, HOOK™
Protein-Extraktion Protein Extraction Kits
The HOOK™ Cell Surface Protein Isolation Kit uses HOOK™ biotin labelling and purification technology in conjunction with Mammalian Cell PE LB™ Lysis Buffer to conveniently label cell surface proteins and isolate them for further analysis, including Western blotting. The Kit is supplied with all the necessary reagents and buffers for convenience and improved reproducibility. The kit is compatible with a wide variety of mammalian cells and can be used to compare treated and untreated cells and differences between different cell lines.
Bestellinformation: Accessories included. Additional supplies available on request, please contact your local Avantor sales office for further details.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge