Fluorescent protease assay, Fluoro™
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-320
786-320EA 587 EUR InStock
786-320
Fluorescent protease assay, Fluoro™
Tests Enzymtests
The Fluoro™ Protease Assay Kit is designed for the quantitative determination of proteases present in a protein sample. The assay uses fluorescein isothiocyanate (FITC)-labelled casein as a general protease substrate. The fluorescein label on the FITC-casein is highly quenched. When the proteases present in the sample of interest digest the FITC-casein substrate into smaller peptides, the quenching of the fluorescence label is relieved and the fluorescence of the substrate is increased. The fluorescence of the FITC-labeled peptide is measured with excitation at 485 nm and emission at 530 nm to determine protease activity. The kit detects picogram level of proteases present in the sample.
The kit is supplied with chemically stabilised MSG-Trypsin™ for use as a general protease control; however, other specific protease standard controls can be used. MSG-Trypsin™ is an ultra-pure trypsin from porcine pancreas, modified by methylation followed by TPCK treatment and is extremely resistant to autolysis. The kit components are sufficient for 1000 assays in a microtitre plate format.
Resorufin-Casein Protease Substrate: A colorimetric substrate that when treated with proteases releases resorufin that has an absorbance of 570 nm (Supplied lyophilised). FITC-Casein Protease Substrate: A fluorescent (fluorescein isothiocyanate (FITC)) substrate that when treated with proteases releases FITC that has an excitation at 485 nm and emission at 535 nm (Supplied lyophilised).
Bestellinformation: Accessories must be ordered separately.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge