Protease Estimation Assay Kit, ProteSEEKER™
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-325
786-325EA 1090 EUR InStock
786-325
Protease Estimation Assay Kit, ProteSEEKER™
Tests Enzymtests
ProteSEEKER™ identifies specific types of proteases with a panel of twelve protease inhibitors and a sensitive colorimetric protease screening assay. ProteSEEKER™ allows researchers to screen their protein samples and establish which specific class of proteases are present and therefore design a highly specific protease inhibitor cocktail using the minimal number of protease inhibitors. Alternatively, ProteSEEKER™ can be used to test existing protease inhibitor cocktails and identify their inadequacies and therefore supplement in additional protease inhibitors.
ProteSEEKER™ protease screening assay consists of a ready-to-use dye-labeled protein, which is digested by proteases to release dye-labeled peptides. The absorbance of which is measured for determination of protease activity. The inhibitors are supplied at a 100X concentration and the 1X concentration provides >90% inhibition in most biological samples.
Bestellinformation: Accessories must be ordered separately.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge