CDI amine reactive resin
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-404
786-404EA 333 EUR InStock
786-404
CDI amine reactive resin
Proteinkonjugate und Markierungen
CDI amine reactive agarose consists of 6% cross-linked agarose activated with CDI (1,1’-carbonyl diimidazole) to form reactive imidazole carbamates.
  • Proven coupling chemistry and stable linkages
  • Easy to use
  • Couple in inorganic buffers for insoluble molecules
  • Couple proteins and peptides
  • Couple primary amine containing ligands
Ideal for immobilising peptides, small organic molecules and certain proteins and reactions can occur in organic solvent making it suitable for water-insoluble ligands.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge