Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Coomassie Brillantblau G 250-Lösung Gel-Elektrophorese-Färbung, anwendungsbereit

Lieferant: G Bioscienes

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


786-497
786-497EA 217 EUR InStock
786-497
Coomassie Brillantblau G 250-Lösung Gel-Elektrophorese-Färbung, anwendungsbereit
Coomassie Brillantblau G 250
G-Biosciences' Coomassie Brilliant Blue (CBB) G-250 and R-250 Stain are based on a colloidal Coomassie stain. CBB G-250 and R-250 stain proteins with high band visibility. The staining of gels with CBB G-250 and R-250 allows the examination of protein bands even during the staining process.

After the staining process, the band intensity may be further enhanced by de-staining the stained gel in our CBB De-Staining Solution or 30% Methanol. CBB G-250 and R-250 Stains have a sensitivity of staining 8-10ng protein/band (e.g., 8-10ng of BSA is visible in 4-20% SDS acrylamide gels). The supplied volume is suitable for 40-50 mini gels (8 x 10cm) & 20-25 large gels (12 x 15cm).

G-Biosciences' Coomassie Brilliant Blue R-250 and G-250 are both offered in dry powder form and convenient ready-to-use solutions.
A destaining solution is also offered.


Formel: C₄₇H₄₈N₃NaO₇S₂
Storage Temperature: Raumtemperatur
MDL Number: MFCD00078482
CAS-Nummer: 6104-58-1
EINECS: 228-058-4
Order Now

 

PRODUKT

TECHNISCHE DATEN

EGÄNZENDE PRODUKTE

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge