IgG Biotinylation kits, HOOK™
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-728 786-729
786-728EA 721 EUR InStock
786-728 786-729
IgG Biotinylation kits, HOOK™
Proteinkonjugate und Markierungen
HOOK™ IgG biotinylation kits are designed for the biotinylation of IgG molecules by immobilising the IgG molecules on a solid support. Nickel chelating resin allows for the rapid removal of uncoupled biotin and therefore eliminates the need for further dialysis or desalting of the biotinylated antibody.
  • Simpler antibody biotinylation
  • Solid support technology eliminates dialysis/desalting
  • Suitable for 1 to 10mg antibody
  • PEG Biotin reagent for reduced steric hindrance and increased labelled antibody solubility
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge