Monoclonal IgG purification kit, Pearl™
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-802
786-802EA 2990 EUR InStock
786-802
Monoclonal IgG purification kit, Pearl™
Protein-Aufbereitungs Protein Purification Kits
The Pearl™ Monoclonal IgG Purification Kit allows for the rapid purification of antibodies from cell culture supernatant and ascites fluid. The Pearl™ IgG purification resin binds the high abundant, non-IgG proteins (i.e. albumin) and allows the IgG molecules to pass through in a physiological buffer. The IgG molecules can be stored or used in downstream applications without further clean-up, such as ammonium sulphate precipitation.
  • One step purification
  • Purification in <15 minutes
  • Faster purification compared to Protein A or Protein G resins
  • >90% recovery with >80% purity
The Pearl™ Monoclonal IgG Purification Kit can be used to purify antibodies direct from cell culture supernatant with less than 10% FBS or can be used with ascites fluid after treatment with the supplied Ascites PreTreat. The kit can purify IgG from ~1L cell culture supernatant or 200 ml ascites fluid.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge