Protein solubilisation buffer
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-PSB
786-PSBEA 348 EUR InStock
786-PSB
Protein solubilisation buffer
Puffer Extraktionspuffer
FOCUS™ protein solubilisation buffer is a dry, urea-based buffer for protein solubilisation. It is a pre-mixed formulation of urea, thiourea, CHAPS and non detergent sulphobetaine for maximum solubilising strength.
  • Convenient and simple to use - simply hydrate and use
  • Prevents urea induced protein carbamylation
  • Prevents waste of unused reagents
Lieferumfang: Supplied with specific dilution buffer for optimal rehydration and protein solubilisation conditions.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge