Supelco® TraceCERT® Organic Reference Standard, Methyl methanesulfonate
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
78697-120MG
78697-120MGEA 80.2 EUR InStock
78697-120MG
Supelco® TraceCERT® Organic Reference Standard, Methyl methanesulfonate
Standards Organische Standards
  • GHS signal: GHS06, GHS08
Zertifizierungen: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Achtung! Bitte beachten: P202 - P273 - P280 - P301 + P310 - P302 + P352 - P305 + P351 + P338
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa4940735c1c0727bf3f72c2ff1f774f8f59c26f&vid=fa4940735c1c0727bf3f72c2ff1f774f8f59c26f&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge