RenLam® M-1 for microscopy, suitable as a embedding medium, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
78711-1L 78711-250ML
78711-1LEA 151 EUR InStock
78711-1L 78711-250ML
RenLam® M-1 for microscopy, suitable as a embedding medium, Sigma-Aldrich®
Färbemittel Mikroskopie- und Färbereagenzien
To be used with SIAL 69571 Ren HY 956 (Hardener for Epoxy Embedding Medium)
Achtung! Bitte beachten: Renlam is a registered trademark of Huntsman Advanced Materials Inc.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge