SDS-PAGE Clean Up Kit
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
80-6484-70
80-6484-70EA 460 EUR InStock
80-6484-70
SDS-PAGE Clean Up Kit
Protein-Aufbereitungs Protein Purification Resins
SDS-PAGE clean up kit is designed to prepare samples for SDS-PAGE that are frequently difficult to analyse due to the presence of salts or low protein concentration.
  • Prepares samples for SDS-PAGE that are otherwise difficult to analyse because of salts or low protein concentrations
  • Quantitatively precipitate proteins while leaving interfering substances in solution
  • Delivers quantitative yield in less than 2 hours
SDS-PAGE clean up kit uses a combination of a precipitant and co-precipitant to quantitatively precipitate the sample proteins while leaving interfering substances in solution. Detergents, salts, lipids, phenolics, and nucleic acids remain soluble while the proteins are pelleted by centrifugation. The precipitate is washed further to remove nonprotein contaminants and centrifuged again. The resultant pellet is resuspended, mixed with SDS-PAGE sample buffer, and heated. The sample is then ready for SDS-PAGE. The procedure can be completed in under 2 hours with quantitative yield.
Bestellinformation: Kit contains sufficient reagents for 50×100 μl samples: 15 ml Precipitant, 15 ml Co-precipitant, 50 ml Wash buffer, 250 μl Wash additive, 2 ml Buffer I, 500 μl Buffer II, Quick reference card and instructions.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge