Isoamylalkohol, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Lieferant: AVANTOR PERFORMANCE MATERIALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
Gefahr
8010.1000EA 91.86 EUR InStock
8010.1000
Isoamylalkohol, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Isoamylalkohol
Formel: (CH₃)₂CHCH₂CH₂OH
Siedepunkt: 130 °C (1013 hPa)
Melting Pt: –117 °C
Dichte: 0,812 g/cm³ (20 °C)
Flash-Pt: 42 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00002934
CAS-Nummer: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3,III
Merck Index: 13,05214
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=60e27777862a1441bdec89c868acf27847ec49b8&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Assay (by GC) min. 98 %
Acids and Esters (as Amyl Acetate) max. 0.06 %
Aldehydes passes test
Boiling Range 128 - 132 °C
Color (APHA) max. 5
Residue after Evaporation max. 0.003 %
Substances Darkened by H2SO4 passes test
Titrable Acid (meq/g) max. 0.002
Water (H2O) max. 0.2 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge