Dry heat indicator tapes, Comply™
Lieferant: 3M MEDICAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M1226/19
804-0226EA 69.4 EUR InStock
804-0226
Dry heat indicator tapes, Comply™
Sterilisationsindikatoren
This chemical indicator tape is designed for use as packaging tape and for the immediate identification of items exposed to dry heat processing.
A self-adhesive, semi-creped paper with an indicator ink printed as stripes across the width of the tape. The tape is white and the ink colour is pale green prior to use.
Verpackung: 48 rolls per box.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge