Benzylalkohol ≥99,9% (durch GC), CHROMASOLV™ für die Headspace-Gaschromatographie, Riedel-de Haën™
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
Achtung
80708-1L
80708-1LEA 271.56 EUR InStock
80708-1L
Benzylalkohol ≥99,9% (durch GC), CHROMASOLV™ für die Headspace-Gaschromatographie, Riedel-de Haën™
Benzylalkohol
Honeywell Chromasolv™ GC-headspace solvents:
  • Developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • Very pure solvents with extremely low concentrations of the defined residual solvents
Formel: C₆H₅CH₂OH
MW: 108.14 g/mol
Siedepunkt: 205.3 °C (1.013 hPa)
Melting Pt: –15.4 °C
Dichte: 1.045 g/cm³ (20 °C)
Flash-Pt: 100.4 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00004599
CAS-Nummer: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=fc6a85fbdb3ccffa1f6db5eed6a9883a84e92c6b&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Assay (GC) Min. 99.9 %
Refractive index (n 20/D) 1.538 - 1.541
Water (Karl Fischer) Max. 0.02 %
Acetophenone (GC) Max. 0.01 %
Benzaldehyde (GC) Max. 0.01 %
Suitability for the GC Headspace complying
Vapor Pressure 3.75 mmHg ( 77 °C)
Autoignition Temperature 817 °F
Vapor Density 3.7 (vs air)
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge