Phytochemical standard, Tanshinone I, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
80714-10MG
80714-10MGEA 143 EUR InStock
80714-10MG
Phytochemical standard, Tanshinone I, analytical standard, Supelco®
Standards Organische Standards
Phenanthrenequinone constituent of Chinese medicinal herb Danshen (Salvia miltiorrhiza). Modulates or prevents breast cancer metastasis by regulating adhesion molecules. Anti-inflammatory.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00d659aca0e14d7e68f691f767fa70fdd9521844&vid=00d659aca0e14d7e68f691f767fa70fdd9521844&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge