Marker, permanent, Sharpie
Lieferant: DIALSTAT OFFICE SUPPLIES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BE4000
811-0023EA 62.7 EUR InStock
811-0023
Marker, permanent, Sharpie
Schreibstifte Permanentmarker
These Sharpie markers with long lasting durable fibre tip are permanent on most hard to mark surfaces.
  • Fine tip line width of 1 mm for precision marking
  • Fade and water resistant
  • Quick-drying ink
Excellent for marking cardboard/photo paper/corrugate/wood/metal and foil.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge