Permanent marker pen, Edding 3000
Lieferant: THEODOR KIHLER BURO EINRICHTUN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
EDD30003
811-3123EA 35.6 EUR InStock
811-3123
Permanent marker pen, Edding 3000
Schreibstifte Permanentmarker
Permananent marker for writing, colouring, labelling for materials such as metal, glass or plastic, low odour ink, round interchangeable tip. Line thickness of 1,5 - 3 mm.
  • Fast drying
  • Waterproof
  • Refillable
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge