Reinraum-Schreibstifte
Lieferant: MICRONOVA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PEN-01IR PEN-02IR PEN-20IR PEN-02 PEN-01 PEN-01TIR PEN-25IR PEN-22IR PEN-51IR PEN-11 PEN-11IR PEN-12 PEN-12IR PEN-23IR PEN-51 PEN-52 PEN-52IR PEN-53IR
818-0550EA 56.3 EUR InStock
818-0550 818-0551 811-0122 818-0244 818-0243 811-0168 811-0174 811-0173 811-0195 811-0192 818-0284 811-0193 818-0285 MICRPEN-23IR MICRPEN-51 MICRPEN-52 811-0196 MICRPEN-53IR
Reinraum-Schreibstifte
Schreibstifte Reinraumstifte
Micronova cleanroom pens feature low sodium, indelible ink and a special low particulate barrel to limit particle generation. Cleanroom irradiated Sharpie® markers write permanently on most surfaces with quick drying, non toxic, alcohol-based ink that is fade and water resistant to further extend multiple project uses. Marks will not smear on reusable glass, metal, or porcelain lab ware, but can be scrubbed off.
  • Indelible, low sodium ink pens
  • White, high density polypropylene barrel
  • Holds up to wipe down with IPA, bleach, and phenols; integrity of printed barrel will be compromised with thinners and strong solvents
  • Available capped or retractable
  • Choice of black, blue or red ink
  • Available gamma irradiated for aseptic environments
Designed for documentation and labelling in cleanrooms and aseptic environments. Gamma irradiated pens and markers are suitable for Aseptic ISO 5 and other clean production areas. The unique polypropylene barrel is manufactured to limit particle generation and holds up to IPA, bleach, and phenol wipe downs. Available irradiated, capped, or click versions, with black, blue, or red ink.
Verpackung: Packed 10 pens per pack and 10 packs per case. Markers available in original capped and retractable styles are individually wrapped and double-bagged with four markers per pack and three packs per case.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge