Cleanroom wipes, Quiltec® I
Lieferant: CONTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
QT1-99 QT1-1212 LWQT0019 QT1-99B QT1-1212B/100 LWQT0034 QT1-66B
115-0378EA 115 EUR InStock
115-0378 115-3077 115-3075 818-0301 115-3078 115-3076 115-3079
Cleanroom wipes, Quiltec® I
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
Quiltec I wipes offer the highest sorptive capacity of any sealed edge wipe available. They are manufactured using two layers of knitted polyester fabric that are laminated together to form a stronger, more adsorbent wipe.
  • Most sorbent sealed edge wipe available
  • Extremely low in both particles and fibres reducing the possibility of contamination
  • Strong fabric is abrasion and chemical resistant
  • Laser-cut, which seals fibres at wipe's edge
  • Excellent sorbency with solvents
  • Laundered and packed in a Class 4 cleanroom
Additional fluid is captured within the quilted pockets between the fabric layers, increasing the sorbent capacity. Quiltec I has more than twice the adsorbent capacity of a single layer wipe.
Recommended for ISO Class 3 - 5 environments.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge