ESD cellulose tape, plastic core, Wescorp
Lieferant: DESCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
242270 242271 242272 242275 242277
819-0143EA 9.6 EUR InStock
819-0143 819-0144 819-0145 819-0146 819-0147
ESD cellulose tape, plastic core, Wescorp
Klebebänder
Wescorp ESD cellulose tape, plastic core are used for general purpose ESD tape applications.
  • Prevents damage to sensitive electronic components in manufacturing
  • Secure (bundle) IC DIP tubes
  • Use with ESD symbols and/or yellow stripe for ESD awareness
  • Lead-free RoHS compliant
Ideal in packaging for container sealing, static shielding bag closure, and holding DIP tubes. Ideal for conformal coating or holding and sealing supplies in manufacturing.
Ideal for holding notes, work orders or instructions in offices, antistatic worksurfaces, or for general purpose third hand use.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge