Silikongummischlaufen
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F203080000
819-1416EA 182 EUR InStock
819-1416
Silikongummischlaufen
Kabelbinder
These stretchy silicone rubber bands offer simple and convenient solutions for your tying, fastening and color coding requirements in the lab and workplace.
  • Adjustable in size
  • Retain elasticity from −49 to 121 ° C
  • 5 mm wide loops can stretch up to 100 mm in diameter
  • Colours: red, blue and yellow
Perfect for use in the incubator, autoclave or freezer and for holding columns and burettes firmly to supports. They can also hold foil covers on flasks, close dialysis bags or biohazard bags and fasten coils of flexible tubing.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge