Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Safranin O für die Mikroskopie, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

M-Clarity™ Qualitätsstufe = MQ200

84120-100G 84120-25G
84120-100GEA 132 EUR InStock
84120-100G 84120-25G
Safranin O für die Mikroskopie, Sigma-Aldrich®
Safranin O
Safranin O, also known as basic red 2, is a basic, cationic, and metachromatic azine dye used as a biological stain. The characteristic azine chromophore contains the phenazine, a planar, tricyclic dibenzopyrazine, which consists of two non-planar benzene rings linked by two nitrogen atoms. A delocalised positive charge, an alternation of aromatic and quinonoid structures, and diazotisable amine groups constitute the chromophoric system. c-Methylation makes this dye of moderate overall size. The trimethyl cationic form of safranine is lipophilic, whereas the dimethyl forms are weakly hydrophilic. The dye is commercially available as a chloride.

  • Safranin O is widely employed as a microbiological stain, especially as a counterstain in gram staining to distinguish gram-positive from gram-negative micro-organisms, and for staining bacterial spores
  • It is used to demonstrate glycosaminoglycans in human and animal histological specimens and nuclei
  • It is a component of Benda's polychrome stain
  • It is a component of various polychromes and lignin stains for botanical staining applications
  • It has also been applied for the demonstration of mast cells in cytospin preparations and staining surgical frozen sections
  • It is a diagnostic assay manufacturing, hematology, histology, microbiology


Formel: C₂₀H₁₉ClN₄
Dichte: 1,447 g/cm³ (22 °C)
Storage Temperature: Room Temperature
MDL Number: MFCD00011759
CAS-Nummer: 477-73-6
EINECS: 207-518-8
ADR: 1,
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=208ff91bb334b473e80398f3a7c448b72f73efbc&vid=208ff91bb334b473e80398f3a7c448b72f73efbc&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (Colour) Green to very dark green
Appearance (Form) Powder or crystals
Thin layer chromatography Corresponds to standard
Solubility (Colour) Red to very dark red
Solubility (turbidity) Clear to turbid
Solubility (method) 0.01 g in 10 ml methanol
pH-transition 1
Colour Red to pink
Wavelength (1) (UV) 525.0 - 535.0 nm
Extinction (1%/1 cm) (1) (uv) ≥ 1000
Solvent (UV) ETOH 50 %

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge